Fisherbrand™ Labeling Tape Edge
Edge
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
 View more versions of this product
Catalog No. 15950
Edge
Explore available promotions
Description
Description
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
  • Strong, durable and oil-, water- and acid-resistant
  • Withstands temperature from -23° to +121°C (-10° to +250°F)
  • Core diameter: 1 in.
  • Can be written on with pen, pencil, or permanent marker
  • Will stick to most surfaces including glass, plastic, and metal
Specifications
Specifications
White
Label Tape
1 Ea.
-23°C to +121°C
14 yd.
2 in.
2.5 cm
Tape
Waterproof, Oil and Acid resistant
-10 to 250°F
14 yd. x 2 in.
13 m
1 in.