Fisherbrand™ Lead-Free Autoclave Tape Edge
Edge
Stripes change to black after sterilization process.
 View more versions of this product
Catalog No. 15-901-110
Edge
Explore available promotions
Description
Description
  • Black stripes appear after exposure to steam sterilization condition
  • Sticks securely under high-temperature and high-pressure conditions
  • Removes easily without leaving sticky residue behind
  • Can be used in temperatures ranging from -23° to +121°C (-9° to +250°F)
  • Available in various sizes
  • Lead-free material
Specifications
Specifications
Self-adhesive
Tape
4 Rolls/Cs.
-23°C to +121°C
54.9 m
White
Steam Sterilization
-9 to 250°F
180 ft.
0.75 in.