Fisherbrand™ Low Form 24-Hour Urine Collection Containers
Low form containers are available with and without pour spout.
Supplier:  Fisherbrand™ ES126911
 View more versions of this product
Catalog No. 22-026-361
Explore available promotions
Description
Description
  • Medical grade High Density Polyethylene (HDPE) container with UV filtering provides safe and easy transport of urine samples and protects light-sensitive specimens
  • Leak-resistant HDPE closure ensures specimen integrity
  • 3.5L with 500mL graduations to assist in sample measurement
  • Ergonomic design allows for easy storage and use by patient and healthcare professionals
  • Available with an easy to use pour spout to aid in specimen testing
  • Centrally located handle for easy use
  • Metal-, zinc-, and latex-free
  • Resistant to weak acids (hydrochloric and acetic acid)
Specifications
Specifications
24-Hour Urine Specimen Collection Container
HDPE
Horizontal Space Saver
70 mm leak resistant HDPE closure with gasketted seal
40/Cs.
Screw Cap
Amber
500 mL
Yes
3.5 L