Fisherbrand™ Low-Nitrogen Weighing Paper Edge
Edge
For analytical balances
 View more versions of this product
Catalog No. 09-898-12B
Edge
Explore available promotions
Description
Description
For analytical balances
  • Sufficient body to support sample
  • Smooth textured to avoid trapping material
  • Does not absorb moisture
Specifications
Specifications
4 x 4 in. (100 x 100mm)
4 in.
For analytical balances
100 mm
4 in.
Low Nitrogen Paper
100 mm
Square