Fisherbrand™ Low-Nitrogen Weighing Paper
For analytical balances
$26.45 - $84.14
Specifications
Product Type Low Nitrogen Paper
Shape Square
For Use With (Application) For analytical balances
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
09-898-12A Edge
View Documents
N/A 3 x 3 in. (76 x 76mm)
Pack of 500 Each for $26.45
Case of 12 Pack for $317.40
Edge
09-898-12B Edge
View Documents
N/A 4 x 4 in. (100 x 100mm)
Pack of 500 Each for $40.28
Case of 12 Pack for $483.36
Edge
09-898-12C Edge
View Documents
N/A 6 x 6 in. (152 x 152mm)
Pack of 500 Each for $84.14
Case of 12 Pack for $1,009.68
Edge
Description
Description
For analytical balances
  • Sufficient body to support sample
  • Smooth textured to avoid trapping material
  • Does not absorb moisture
Specifications
Specifications
Low Nitrogen Paper
Square
For analytical balances
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'