Fisherbrand™ Low-Retention Microcentrifuge Tubes
Provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.
 View more versions of this product
Catalog No. 02-681-320
Explore available promotions
Description
Description
Snap-Cap™ lid provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.
  • Manufactured from durable chemical-resistant polypropylene
  • 0.6mL tube is graduated in 0.2mL increments; 1.5mL tube, in 0.5mL increments with an additional graduation at 0.1 and 2.0mL size, in 0.5mL increments
  • Designed for use in critical applications where minute sample recovery is important
  • Withstand speeds up to 30,000xG
  • Recommended for freezer-to-centrifuge use
  • Autoclavable up to 120°C and withstand freezing temperatures as low as -80°C
Specifications
Specifications
Snap Cap
1.5 mL
40 mm
Critical applications where minute sample recovery is important
Non-sterile
Snap Cap
13 mm
Autoclavable
Natural
Polypropylene
Bulk
Yes
30,000 x g
Videos
SDS
Documents
Documents
Accessories