missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Microcentrifuge Tubes

Provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.

$64.85 - $214.15

Specifications

Cap Style Snap Cap
Autoclavable Autoclavable
Color Natural
Material Polypropylene
For Use With (Application) Critical applications where minute sample recovery is important
View More Specs

Products 5
Catalog Number Mfr. No. Closure Type Capacity (Metric) Sterility Price Quantity  
Catalog Number Mfr. No. Closure Type Capacity (Metric) Sterility Price Quantity  
02-681-311
Accessories View Documents
N/A Snap Cap 0.6 mL Non-sterile
Pack of 500 Each for $86.74
Case of 20 Pack for $1,734.80
 
02-681-320
Accessories View Documents
N/A Snap Cap 1.5 mL Non-sterile
Pack of 250 Each for $64.85
Case of 20 Pack for $1,297.00
 
02-681-321
Accessories View Documents
N/A Snap Cap 2 mL Non-sterile
Pack of 250 Each for $78.33
Case of 20 Pack for $1,566.60
 
02-681-331
Accessories View Documents
N/A Attached Cap 1.5 mL Sterile
Pack of 250 Each for $122.70
Case of 10 Pack for $1,227.00
 
02-681-332
Accessories View Documents
N/A Attached Cap 2 mL Sterile
Pack of 250 Each for $214.15
Case of 10 Pack for $2,141.50
 
Description

Description

Snap-Cap™ lid provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.

  • Manufactured from durable chemical-resistant polypropylene
  • 0.6mL tube is graduated in 0.2mL increments; 1.5mL tube, in 0.5mL increments with an additional graduation at 0.1 and 2.0mL size, in 0.5mL increments
  • Designed for use in critical applications where minute sample recovery is important
  • Withstand speeds up to 30,000xG
  • Recommended for freezer-to-centrifuge use
  • Autoclavable up to 120°C and withstand freezing temperatures as low as -80°C
  • RNase/DNase free

Order Info

Nonsterile tubes supplied in bulk pack. Sterile tubes (natural colored) come individually wrapped.

Specifications

Specifications

Snap Cap
Natural
Critical applications where minute sample recovery is important
30,000 x g
Autoclavable
Polypropylene
Yes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'