Fisherbrand™ Manual Differential Counter
Designed for blood cell counting
Supplier:  Fisherbrand™ 505CIF
 View more versions of this product
Catalog No. 13-684-140
Explore available promotions
Includes: Stip chart
Description
Description
Designed for blood cell counting
  • Helps in automatic calculation including percentages
  • Totaling unit rings a bell at 100 increments
  • Dual reset knobs
  • Last unit is the totaling unit, which records total number of strokes made on other keys
  • Removable tabs on top of counter provide area for identification of classification being counted
  • Acid resistant, non slip bottom
Specifications
Specifications
Differential Cell Counter
5
Stip chart
0 to 999
Blood Cell Counting