Fisherbrand™ Manual Differential Counter
Designed for blood cell counting
$243.77 - $277.00
Specifications
Product Type | Differential Cell Counter |
---|---|
Displayed Text | 0 to 999 |
For Use With (Application) | Blood Cell Counting |
Includes | Stip chart |
Includes: Stip chart
Catalog Number | Mfr. No. | No. of Keys | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Keys | Price | Quantity | |||||
13-684-140
|
Fisherbrand™
505CIF |
5 |
Each for $243.77
|
|
|||||
13-684-141
|
Fisherbrand™
808CIF |
8 |
Each for $277.00
|
|
|||||
Description
Designed for blood cell counting
- Helps in automatic calculation including percentages
- Totaling unit rings a bell at 100 increments
- Dual reset knobs
- Last unit is the totaling unit, which records total number of strokes made on other keys
- Removable tabs on top of counter provide area for identification of classification being counted
- Acid resistant, non slip bottom
Specifications
Differential Cell Counter | |
0 to 999 | |
Blood Cell Counting | |
Stip chart |