Fisherbrand™ Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
 View more versions of this product
Catalog No. 14-955-118A
Explore available promotions
Description
Description
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable
  • Container is polypropylene; Cap is LDPE
Specifications
Specifications
Specimen Container
Tall, Thin
32.0 oz.
LDPE
11.44 cm
13.32 cm
Polypropylene
Not Autoclavable
946 mL
Translucent
5.24 in.