Fisherbrand™ Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
$95.65 - $333.00
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Material | Color | Shape | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Material | Color | Shape | Price | Quantity | ||
14-955-114A
|
N/A | 118 mL | Polypropylene | Translucent | Short, Wide |
Case of 300 Each for $259.00
|
|
||
14-955-114B
|
N/A | 148 mL | Polypropylene | Translucent | Tall, Thin |
Case of 300 Each for $333.00
|
|
||
14-955-115A
|
N/A | 237 mL | LDPE | Translucent | Short, Wide |
Case of 100 Each for $142.30
|
|
||
14-955-115B
|
N/A | 237 mL | LDPE | White | Short, Wide |
Case of 100 Each for $144.45
|
|
||
14-955-116A
|
N/A | 237 mL | LDPE | Translucent | Tall, Thin |
Case of 100 Each for $126.40
|
|
||
14-955-116B
|
N/A | 237 mL | LDPE | White | Tall, Thin |
Case of 100 Each for $128.90
|
|
||
14-955-117A
|
N/A | 473 mL | LDPE | Translucent | Short, Wide |
Case of 100 Each for $185.50
|
|
||
14-955-117B
|
N/A | 473 mL | LDPE | White | Tall, Thin |
Case of 100 Each for $206.15
|
|
||
14-955-118A
|
N/A | 946 mL | LDPE | Translucent | Tall, Thin |
Case of 100 Each for $288.00
|
|
||
14-955-118B
|
N/A | 1100 mL | LDPE | White | Short, Wide |
Case of 100 Each for $288.00
|
|
||
14-955-119A
|
N/A | 1900 mL | LDPE | Translucent | Tall, Thin |
Case of 50 Each for $254.00
|
|
||
14-955-119B
|
N/A | 1900 mL | LDPE | White | Tall, Thin |
Case of 50 Each for $254.00
|
|
||
14-955-120A
|
N/A | 2500 mL | LDPE | Translucent | Tall, Thin |
Case of 25 Each for $143.70
|
|
||
14-955-120B
|
N/A | 2500 mL | LDPE | White | Tall, Thin |
Case of 25 Each for $149.10
|
|
||
14-955-121A
![]() |
N/A | 5100 mL | LDPE | Translucent | Short, Wide |
Case of 10 Each for $95.65
|
![]() |
||
14-955-121B
|
N/A | 5100 mL | LDPE | White | Tall, Thin |
Case of 10 Each for $97.40
|
|
||
Description
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
- Resist freezing and boiling
- Stackable for compact storage
- Select containers are autoclavable (see ordering table); lids are not autoclavable
- Short/wide style
- Tall/thin style
- Graduated
Specifications
Specimen Container |