Fisherbrand™ Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
 View more versions of this product
Catalog No. 14-955-119A
Explore available promotions
Description
Description
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable
Specifications
Specifications
Specimen Container
Tall, Thin
1900 mL
Translucent
6.13 in.
HDPE
64.0 oz.
LDPE
14.68 cm
15.58 cm