Fisherbrand™ Multipurpose Specimen Storage Containers Edge
Edge
Inert to most chemicals including formaldehyde, weak acids and all bases
 View more versions of this product
Catalog No. 14-955-121A
Edge
Explore available promotions
Description
Description
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
  • Resist freezing and boiling
  • Stackable for compact storage
  • Select containers are autoclavable (see ordering table); lids are not autoclavable
Specifications
Specifications
Specimen Container
Short, Wide
5100 mL
Translucent
6.98 in.
HDPE
172 oz.
LDPE
22.0 cm
17.72 cm