Fisherbrand™ NFR Self-Laminating Labels
For use on bottles, drums or any containers holding hazardous substances.
Supplier: Fisherbrand™ 68801
Description
- Each self-adhesive label features a convenient laminate cover and space to fill in National Fire Protection Association (NFPA) hazard ratings
- Add the appropriate hazard rating number and circle the symbols of personal protection required
- Peel off backing to apply
Specifications
Label | |
Bottles, drums or any containers holding hazardous substances | |
3.5 in. | |
5 cm |
NFR Self-Laminating Label | |
25/Pk. | |
2 in. |