missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ NFR Self-Laminating Labels
For use on bottles, drums or any containers holding hazardous substances.
$11.34 - $17.43
Specifications
Format | Label |
---|---|
Product Type | NFR Self-Laminating Label |
For Use With (Application) | Bottles, drums or any containers holding hazardous substances |
Quantity | 25/Pk. |
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Width (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Width (English) | Price | Quantity | |||
18-999-791
|
Fisherbrand™
22681 |
3 in. | 8 cm | 5 in. |
Each for $17.43
|
|
|||
18-999-790
|
Fisherbrand™
68801 |
2 in. | 5 cm | 3.5 in. |
Each for $11.34
|
|
|||
Description
- Each self-adhesive label features a convenient laminate cover and space to fill in National Fire Protection Association (NFPA) hazard ratings
- Add the appropriate hazard rating number and circle the symbols of personal protection required
- Peel off backing to apply
Specifications
Label | |
NFR Self-Laminating Label | |
Bottles, drums or any containers holding hazardous substances | |
25/Pk. |