Fisherbrand™ Nonsterile Plastic Culture Tubes
Extra-heavy wall construction is ideal for numerous testing applications
 View more versions of this product
Catalog No. 14-959-16A
Explore available promotions
Description
Description
Extra-heavy wall construction is ideal for numerous testing applications
  • Round bottom
  • Special molding avoids leaks and cracks
  • 12 O.D. × 75mm long tube has 5mL capacity and heavier wall than competitive tubes
  • Slight lip at top provides seal when used with Cat. No. 14-961-12 Snap Cap (sold separately)
Capacity
  • 12x75mm tubes: 5mL
  • 13x100mm tubes: 8mL
  • 16x125mm tubes: 16mL
  • 17x100mm tubes: 14mL
Order Info
Select 12 x 75mm tubes available with special one-directional, oriented-box packaging (see ordering information). Caps for 12mm and 17mm tubes available separately (14-961-12, 14-961-15).
Specifications
Specifications
Translucent
75 mm
Single-use
1000/Cs.
12 mm
Culture Tube
5 mL
Polypropylene
Ideal for numerous testing applications
Non-sterile
Not Included