Fisherbrand™ Pipet Tips in SureGrip Racks
Tips in sturdy, reusable racks
$166.93 - $172.57
Specifications
Sterility Sterile
Autoclavable Autoclavable
Format Rack
Graduated Yes
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Color Tip Style Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Color Tip Style Price Quantity  
02-100-506
View Documents
N/A 10 μL Natural Standard Length
Pack of 960 Each for $170.96
Case of 5 Pack for $854.80
02-100-507
View Documents
N/A 10 μL Natural Extended Length
Pack of 960 Each for $172.57
Case of 5 Pack for $862.85
02-100-508
View Documents
N/A 200 μL Natural Extended Length
Pack of 960 Each for $167.61
Case of 5 Pack for $838.05
02-100-509
View Documents
N/A 200 μL Yellow Extended Length
Pack of 960 Each for $167.61
Case of 5 Pack for $838.05
02-100-510
View Documents
N/A 1250 μL Natural Extended Length
Pack of 768 Each for $166.93
Case of 5 Pack for $834.65
Description
Description
Pipet Tips in Racks
  • All tips are available racked in sturdy, reusable CellRacks
  • Racks can withstand numerous autoclave cycles
  • Keeps tips aligned for pipetter mounting
Several Tip Sizes Available
  • 10μL Pipet tip features graduation marks at 2μL and 10μL
  • 10μL Long Reach tip features extended length (45mm) that reaches the bottom of tubes without contaminating the pipetter shaft, and graduation marks at 2μL, 5μL and 10μL
  • 200μL Pipet tip features a soft fit collar for easy mounting and eject, graduation marks at 10μL, 50μL and 100μL, and is available in natural and yellow
  • 1250μL Pipet tips feature an extended length (103mm), racked on 9mm centers (multichannel compatible), and graduation marks at 200μL, 500μL and 1000μL
Specifications
Specifications
Sterile
Autoclavable
Rack
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'