Fisherbrand™ Pipette Stand
Accommodate most major brands of pipetters.
 View more versions of this product
Catalog No. 06-667-1
Explore available promotions
Description
Description
Accommodate most major brands of pipetters.
  • One-position stand (06-667-3) has a single insert hole 1.25 in. (31.8mm) in diameter
  • Three-, four-, and six-position stands have both upper (1.03 in. [26.1mm]) and lower slots (0.625 in. [16mm] in diameter)
  • Multichannel stand is clear acrylic with slots 1 in. (25.5mm) in diameter
Specifications
Specifications
Pipet Stand
Clear, Transparent
HDPE
Single-Channel Pipette
Not Autoclavable
Pipettors (4)
4
20.3 x 20.3 x 30.4cm (8 x 8 x 12 in.)