missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 1827B

 View more versions of this product

Catalog No. 01-827B


Explore available promotions

Description

Description

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications

Specifications

OSHA, 29 CFR1910.1030
Clear with Red Print
61 cm
Autoclavable
12 in.
1.50 mil
Autoclavable Bags
Elastic
24 x 12 in. (61 x 30 cm)
30 cm
Printed in English and Spanish
24 in.
Polyethylene
127°C
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions