Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
$35.20 - $368.48
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polyethylene |
Product Type | Autoclavable Bags |
Temperature (Metric) | 127°C |
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness | Price | Quantity | |||
01-814-1
|
Fisherbrand™
18141 |
Natural | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
Case of 200 Each for $61.23
|
|
|||
01-814-3
|
Fisherbrand™
18143 |
Natural | 24 x 36 in. (61 x 91 cm) | 1.50 mil |
Case of 200 Each for $152.14
|
|
|||
01-827A
|
Fisherbrand™
1827A |
Clear with Red Print | 12 x 8 in. (30 x 20 cm) | 1.50 mil |
Case of 200 Each for $35.20
|
|
|||
01-827D
|
Fisherbrand™
1827D |
Clear with Red Print | 36 x 24 in. (91 x 61 cm) | 1.50 mil |
Case of 200 Each for $212.41
|
|
|||
01-827B
|
Fisherbrand™
1827B |
Clear with Red Print | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
Case of 200 Each for $86.00
|
|
|||
01-827C
|
Fisherbrand™
1827C |
Clear with Red Print | 30 x 24 in. (76 x 61 cm) | 1.50 mil |
Case of 200 Each for $179.17
|
|
|||
01-829A
|
Fisherbrand™
1829A |
Red with Black Print | 12 x 8 in. (30 x 20 cm) | 1.50 mil |
Case of 200 Each for $36.71
|
|
|||
01-829B
|
Fisherbrand™
1829B |
Red with Black Print | 19 x 14 in. (48 x 36 cm) | 1.50 mil |
Case of 200 Each for $82.27
|
|
|||
01-829C
|
Fisherbrand™
1829C |
Red with Black Print | 23 x 19 in. (58 x 48 cm) | 1.50 mil |
Case of 200 Each for $123.22
|
|
|||
01-829D
|
Fisherbrand™
1829D |
Red with Black Print | 25 x 35 in. (64 x 89 cm) | 1.50 mil |
Case of 200 Each for $214.42
|
|
|||
01-829F
|
Fisherbrand™
1829F |
Red with Black Print | 30 x 33 in. (76 x 84 cm) | 1.50 mil |
Case of 200 Each for $235.76
|
|
|||
01-831A
|
Fisherbrand™
1831A |
Red with Black Print | 36 x 24 in. (91 x 61 cm) | 4 mil |
Case of 100 Each for $368.48
|
|
|||
01-832A
|
Fisherbrand™
1832A |
Clear | 36 x 24 in. (91 x 61 cm) | 4 mil |
Case of 100 Each for $228.46
|
|
|||
01-814-2
|
Fisherbrand™
18142 |
Clear | 30 x 24 in. (76 x 61 cm) | 1.50 mil |
Case of 200 Each for $150.23
|
|
|||
01-829E
|
Fisherbrand™
1829E |
Red with Black Print | 48 x 38 in. (122 x 97 cm) | 1.50 mil |
Case of 100 Each for $203.18
|
|
|||
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
Autoclavable | |
Polyethylene | |
Autoclavable Bags | |
127°C |