missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Supplier: Fisherbrand™ 1827C
Description
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
OSHA, 29 CFR1910.1030 | |
Clear with Red Print | |
76 cm | |
Autoclavable | |
24 in. | |
1.50 mil | |
Autoclavable Bags |
Elastic | |
30 x 24 in. (76 x 61 cm) | |
61 cm | |
Printed in English and Spanish | |
30 in. | |
Polyethylene | |
127°C |