Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Supplier:  Fisherbrand™ 44560
 View more versions of this product
Catalog No. 22044560
Explore available promotions
Description
Description
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Specifications
Twist Tie
36 x 24 in. (91 x 61 cm)
61 cm
Biohazard symbol and text in English and Spanish
36 in.
Polyethylene
Clear with Red Print
91 cm
Autoclavable
24 in.
1.50 mil