missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
$24.04 - $122.58
Specifications
Closure Type | Twist Tie |
---|---|
Autoclavable | Autoclavable |
Includes | Biohazard symbol and text in English and Spanish |
Material | Polyethylene |
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Thickness | Price | Quantity | |||
22044559
|
Fisherbrand™
44559 |
Clear with Red Print | 30 x 24 in. (76 x 61 cm) | 1.50 mil |
Case of 100 Each for $51.52
|
|
|||
22044558
|
Fisherbrand™
44558 |
Clear with Red Print | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
Case of 100 Each for $24.04
|
|
|||
22044561
|
Fisherbrand™
44561 |
Red with Black Print | 12 x 8 in. (30 x 20 cm) | 1.25 mil |
Case of 1000 Each for $81.10
|
|
|||
22-044562
|
Fisherbrand™
44562 |
Red with Black Print | 19 x 14 in. (48 x 36 cm) | 1.25 mil |
Case of 200 Each for $43.50
|
|
|||
22044560
|
Fisherbrand™
44560 |
Clear with Red Print | 36 x 24 in. (91 x 61 cm) | 1.50 mil |
Case of 100 Each for $57.42
|
|
|||
22-044563
|
Fisherbrand™
44563 |
Red with Black Print | 23 x 19 in. (58 x 48 cm) | 1.25 mil |
Case of 200 Each for $66.10
|
|
|||
22044564
|
Fisherbrand™
44564 |
Red with Black Print | 35 x 25 in. (89 x 64 cm) | 1.50 mil |
Case of 200 Each for $122.58
|
|
|||
22044565
|
Fisherbrand™
44565 |
Red with Black Print | 36 x 45 in. (91 x 114 cm) | 1.50 mil |
Case of 100 Each for $103.65
|
|
|||
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Twist Tie | |
Autoclavable | |
Biohazard symbol and text in English and Spanish | |
Polyethylene |