Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Supplier:  Fisherbrand™ 44562
 View more versions of this product
Catalog No. 22-044562
Explore available promotions
Description
Description
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Specifications
Twist Tie
19 x 14 in. (48 x 36 cm)
36 cm
Biohazard symbol and text in English and Spanish
19 in.
Polyethylene
Red with Black Print
48 cm
Autoclavable
14 in.
1.25 mil