Fisherbrand™ Polygon Stir Bars
Stir bars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
$3.43 - $18.25
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Alnico Magnet |
Quantity | 50 |
Shape | Polygon |
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | ||
14-512-124
|
N/A | 0.3125 in. | 8 mm | 0.5 in. | 12.7 mm |
Each for $5.89
Case of 50 Each for $294.50
|
|
||
14-512-125
|
N/A | 0.3125 in. | 8 mm | 0.78 in. | 20 mm |
Each for $5.62
Case of 50 Each for $281.00
|
|
||
14-512-126
![]() |
Fisherbrand™
371205038 |
0.375 in. | 9.5 mm | 1 in. | 25.4 mm |
Each for $3.43
Case of 50 Each for $171.50
|
![]() |
||
14-512-127
|
N/A | 0.3125 in. | 8 mm | 2 in. | 50.8 mm |
Each for $9.04
Case of 50 Each for $452.00
|
|
||
14-512-128
|
N/A | 0.375 in. | 10 mm | 2.7 in. | 70 mm |
Each for $18.25
Case of 50 Each for $912.50
|
|
||
16800507
|
Fisherbrand™
371205010 |
0.25 in. | 6 mm | 0.375 in. | 10 mm |
Each for $5.53
Case of 50 Each for $276.50
|
|
||
16800508
|
Fisherbrand™
371205015 |
0.25 in. | 6 mm | 0.625 in. | 15 mm |
Each for $5.27
Case of 50 Each for $263.50
|
|
||
16800509
|
Fisherbrand™
371205025 |
0.3125 in. | 8 mm | 1 in. | 25 mm |
Each for $6.37
Case of 50 Each for $318.50
|
|
||
16800510
|
Fisherbrand™
371205030 |
0.3125 in. | 8 mm | 1.1875 in. | 30 mm |
Each for $6.80
Case of 50 Each for $340.00
|
|
||
16800511
|
Fisherbrand™
371205040 |
0.3125 in. | 8 mm | 1.625 in. | 40 mm |
Each for $7.63
Case of 50 Each for $381.50
|
|
||
16800512
|
Fisherbrand™
371205060 |
0.25 in. | 7 mm | 2.3125 in. | 60 mm |
Each for $8.47
Case of 50 Each for $423.50
|
|
||
16800513
|
Fisherbrand™
371205080 |
0.375 in. | 10 mm | 3.125 in. | 80 mm |
Each for $14.90
Case of 50 Each for $745.00
|
|
||
Description
Stir bars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable | |
Alnico Magnet | |
50 | |
Polygon |