Fisherbrand™ Polypropylene Graduated Specimen Containers
Ideal for specimen collection, transportation and storage
Description
- Autoclavable
- Graduated
- With or without nonautoclavable polyethylene snap lid
Order Info
Note: Sterile containers with lids are individually wrapped, and are sterile inside container.
Specifications
Yes | |
Polypropylene | |
5 oz. |
Yes | |
Sterile | |
148 mL |