Fisherbrand™ Polypropylene Graduated Specimen Containers
Ideal for specimen collection, transportation and storage
$159.95 - $529.00
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Included | Sterility | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Included | Sterility | Price | Quantity | ||
14-955-101
|
N/A | 4.5 oz. | 133 mL | No | Sterile |
N/A
|
|
||
14-955-102
|
N/A | 5 oz. | 148 mL | Yes | Sterile |
N/A
|
|
||
14-955-103
|
N/A | 4.5 oz. | 133 mL | No | Non-sterile |
N/A
|
|
||
14-955-104
|
N/A | 8 oz. | 237 mL | No | Sterile |
N/A
|
|
||
14-955-105
|
N/A | 8 oz. | 237 mL | Yes | Sterile |
N/A
|
|
||
14-955-106
|
N/A | 8 oz. | 237 mL | No | Non-sterile |
N/A
|
|
||
Description
- Autoclavable
- Graduated
- With or without nonautoclavable polyethylene snap lid
Order Info
Note: Sterile containers with lids are individually wrapped, and are sterile inside container. Nonsterile and sterile containers without lids are bulk packed.
Specifications
Yes | |
Polypropylene |