Fisherbrand™ Quartz Ultramicro cells
Available with a variety of window materials for wide range of assays
Supplier: Fisherbrand™ 105202QSZ15
Includes: Fitted PTFE covers or tapered PTFE stoppers
Description
- 8.5mm center height fits most Beckman™ Instruments and Spectronic™ spectrophotometers
- 15.0mm cells fit most Hewlett-Packard™, Perkin-Elmer™, Shimadzu™, Unicam™ and GBC spectrophotometers Choice of correct center height is made such that optical aperture matches center position of light beam
Specifications
50 μL | |
Quartz | |
15 mm |