Fisherbrand™ Quartz Ultramicro cells
Available with a variety of window materials for wide range of assays
$1626.32 - $1626.32
Specifications
Material | Quartz |
---|
Includes: Fitted PTFE covers or tapered PTFE stoppers
Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity | ||||
14-385-928D
|
Fisherbrand™
105201QS10Z15 |
100 μL | 15 mm |
Each for $1,626.32
|
|
||||
14-385-928E
|
Fisherbrand™
105202QSZ85 |
50 μL | 8.5 mm |
Each for $1,626.32
|
|
||||
14-385-928F
|
Fisherbrand™
105202QSZ15 |
50 μL | 15 mm |
Each for $1,626.32
|
|
||||
Description
- 8.5mm center height fits most Beckman™ Instruments and Spectronic™ spectrophotometers
- 15.0mm cells fit most Hewlett-Packard™, Perkin-Elmer™, Shimadzu™, Unicam™ and GBC spectrophotometers Choice of correct center height is made such that optical aperture matches center position of light beam
Specifications
Quartz |