Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Single graduation ring blasted on the neck
 View more versions of this product
Catalog No. 10-205E
Explore available promotions
Includes:
  • 41900R ground glass stopper with marking spot
Description
Description
Single graduation ring blasted on the neck
  • Calibrated “To Contain”
  • Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
  • Do not heated above 150°C when dry
Compliance
  • Designed from ASTM™ Specification E 288, Class A unserialized requirementsr
Specifications
Specifications
Calibrated To Contain
500 mL
1
Yes
Glass
Glass
±0.2 mL
19
Videos
SDS
Documents
Documents
Accessories