Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Single graduation ring blasted on the neck
$88.97 - $771.69
Specifications
Calibration | Calibrated To Contain |
---|---|
Closure Material | Glass |
Material | Glass |
No. per Case | 1 |
Graduated | Yes |
Includes:
- 41900R ground glass stopper with marking spot
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
FB4002000
|
N/A | 2000 mL | 27 |
Each for $189.10
Case of 4 Each for $756.40
|
|
||||
FB40010
|
N/A | 10 mL | 9 |
Case of 12 Each for $771.69
|
|
||||
FB400200
|
N/A | 200 mL | 16 |
Pack of 6 Each for $547.86
Case of 2 Pack for $1,095.72
|
|
||||
10-205A
|
N/A | 25 mL | 9 |
Each for $88.97
Case of 12 Each for $1,067.64
|
|
||||
10-205B
|
N/A | 50 mL | 9 |
Each for $100.62
Case of 12 Each for $1,207.44
|
|
||||
10-205C
|
N/A | 100 mL | 13 |
Each for $107.14
Case of 12 Each for $1,285.68
|
|
||||
10-205D
|
N/A | 250 mL | 16 |
Each for $143.41
Case of 12 Each for $1,720.92
|
|
||||
10-205E
|
N/A | 500 mL | 19 |
Each for $193.72
Case of 12 Each for $2,324.64
|
|
||||
10-205F
|
N/A | 1000 mL | 22 |
Each for $235.31
Case of 6 Each for $1,411.86
|
|
||||
Description
Single graduation ring blasted on the neck
- Calibrated “To Contain”
- Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
- Do not heated above 150°C when dry
Compliance
- Designed from ASTM™ Specification E 288, Class A unserialized requirementsr
Specifications
Calibrated To Contain | |
Glass | |
Yes |
Glass | |
1 |