Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper
Single graduation ring blasted on the neck
Includes:
- 41900R ground glass stopper with marking spot
Description
Single graduation ring blasted on the neck
- Calibrated “To Contain”
- Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
- Do not heated above 150°C when dry
Compliance
- Designed from ASTM™ Specification E 288, Class A unserialized requirementsr
Specifications
Calibrated To Contain | |
10 mL | |
1 | |
9 |
Glass | |
Glass | |
Yes |