Fisherbrand™ Reusable Glass Heavy-Duty Low-Form Beakers
Spout designed for optimum performance
 View more versions of this product
Catalog No. FB1011000
Explore available promotions
Alert:
Not recommended for use on hotplates where significant thermal gradients exist due to lack of agitation of contents or absence of a temperature control.
Description
Description
Fisherbrand™ Reusable Glass Heavy-Duty Low-Form Beakers feature a sturdy construction ideal for mechanical shock and other harsh applications. Spout is designed for optimum performance.
  • Thick uniform walls
  • Evenly tooled top rim with extra wall thickness
  • Durable matte finish marking area
  • White graduated scale
  • Spout designed for optimum performance
Specifications
Specifications
±5%
4.21 in.
1000 mL
Offer Superior Mechanical Strength and Durability, Provides Improved Safety When Used Under Extreme Conditions Such as Mechanized Washing and Rough Handling
5.98 in.
Spout
Matte Finish Marking
Beaker
Yes
Dual Scale
33.81 oz.
10.7 cm
Borosilicate Glass
100 to 1000 mL
15.2 cm
50 mL
6 Pk.
Reusable
ASTM E960, ASTM E438, USP Type I
Videos
SDS
Documents
Documents
Accessories