Fisherbrand™ Reusable Polyethylene Ladles
Extra-heavy-duty for sampling and dispensing corrosive liquids.
 View more versions of this product
Catalog No. 14-242-40
Explore available promotions
Description
Description
Extra-heavy-duty for sampling and dispensing corrosive liquids.
  • Linear polyethylene beaker
  • Secured with welded solid handle
Order Info
Additional ladle sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
Ladle
20.3 oz.
For sampling and dispensing corrosive liquid
13 in.
5.03 in.
12.77 cm
4 in.
HDPE
600 mL
5.03 x 4 x 13 in. (12.77 x 10.16 x 33.02 cm)
33.02 cm
10 in.
25.4 cm
10.16 cm