Fisherbrand™ Reusable Polyethylene Ladles
Extra-heavy-duty for sampling and dispensing corrosive liquids.
$29.52 - $100.41
Specifications
Product Type | Ladle |
---|---|
Material | HDPE |
For Use With (Application) | For sampling and dispensing corrosive liquid |
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity | ||||
14-242-20
|
N/A | 50 mL | 2.5 x 2 x 10.5 in. (6.35 x 5.08 x 26.67 cm) |
Each for $29.52
Case of 12 Each for $354.24
|
|
||||
14-242-25
|
N/A | 100 mL | 3 x 2.3 x 10.75 in. (7.6 x 5.84 x 27.3 cm) |
Each for $32.90
Case of 12 Each for $394.80
|
|
||||
14-242-30
|
N/A | 250 mL | 4.5 x 3.3 x 12.43 in. (11.43 x 8.38 x 31.57 cm) |
Each for $40.55
Case of 12 Each for $486.60
|
|
||||
14-242-40
|
N/A | 600 mL | 5.03 x 4 x 13 in. (12.77 x 10.16 x 33.02 cm) |
Each for $45.90
Case of 12 Each for $550.80
|
|
||||
14-242-45
|
N/A | 1000 mL | 6.62 x 5.31 x 16.12 in. (16.81 x 13.48 x 40.94 cm) |
Each for $100.41
Case of 6 Each for $602.46
|
|
||||
Description
Extra-heavy-duty for sampling and dispensing corrosive liquids.
- Linear polyethylene beaker
- Secured with welded solid handle
Order Info
Additional ladle sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Ladle | |
HDPE | |
For sampling and dispensing corrosive liquid |