Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
Description
- Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Thermo Hydrometer | |
0 to 35° | |
For determining the percentage by weight of sucrose |
NIST | |
Lead and Mercury Free |