Fisherbrand™ Self-Adhesive Label Tape
All-purpose—even sticks to plastics during autoclaving.
 View more versions of this product
Catalog No. 15938
Explore available promotions
Includes: This is a Kit with Label Tape and Dispenser.
Description
Description
All-purpose—even sticks to plastics during autoclaving
  • Sticks to glass, plastic, paper, and wood from -23° to +121°C (-9° to +250°F)
  • Especially suited for use in autoclave, refrigerator, deep freeze, sterilizer and incubator
  • Easily marked with pencil, pen, ballpoint and most felt-tip markers
  • Removes without residue
  • Oil, water and acid resistant
Specifications
Specifications
Self-adhesive
Tape
Autoclave, Refrigerator/freezer, Sterilizer, Incubator
1 Ea.
-23°C to +121°C
14 yd.
0.75 in.
White
Self Adhesive Label Tape
Tape and Dispenser
-9 to 250°F
14 yd. x 0.75 in.
13 m
1 in. I.D.