Fisherbrand™ Serialized/Certified Measuring (Mohr) Pipets
Reusable, Class A pipets with a permanently marked serial number.
 View more versions of this product
Catalog No. 13-665-3M
Explore available promotions
Description
Description
Reusable, Class A pipets with a permanently marked serial number.
  • Fast, uniform flow times
  • Tuf-Temp™-treated to reduce chipping
  • Permanent amber markings are fused to borosilicate glass
  • Accuracy printed on each pipet
  • A total of 23 characters, including spaces, can be imprinted on the pipet
Order Info
Pipets are also available as PersonaPet™ personalized pipets.
Certifications
Include a certificate indicating that calibration is traceable to standards provided by NIST™.
Compliance
Class A, Type 1 pipets are calibrated To Deliver. Meet requirements of Fed. Spec. NNN-P-350C, Style 1-Standard Taper, and ASTM™ E1293.
Specifications
Specifications
±0.03 mL
A
Yes
Permanent Amber marking
Orange
Graduated Pipet, Type 1
NIST, NNN-P-350C, ASTM E1293
Borosilicate Glass
1/10 mL
0.33 oz.
10 mL
Reusable
Product Suggestions
Product Suggestions