Fisherbrand™ Serialized Class A Cylinders with Certificate of Calibration
Calibrated To Deliver.
 View more versions of this product
Catalog No. 08-557HC
Explore available promotions
Includes: Individual fused-in serial numbers and, in addition to traceability certification, are supplied with Certificates of Calibration that list apparent volumes and calibration correction factors
Description
Description
Calibrated To Deliver.
  • Traceability
  • White scale
Specifications
Specifications
Round
Graduated Cylinder
To Deliver
Hexagonal
520 mm
2000 mL
20 mL
±6mL
±2 mL
20.4 in.
White
A
Borosilicate Glass
Individual Fused-In Serial Numbers, Traceable NIST Certificate
Autoclavable
20mL
Videos
SDS
Documents
Documents
Accessories