Fisherbrand™ Shaker Flasks
Reduce the risk of cross-contamination.
 View more versions of this product
Catalog No. BBV250
Explore available promotions
Description
Description
Reduce the risk of cross-contamination.
  • Nonpyrogenic and noncytotoxic
  • Clear, durable polycarbonate with molded-in graduations
  • Polypropylene closure with 0.22μm PTFE pore
  • Individually packed for easy storage and handling
  • Plain bottom for use on benchtop or baffled bottom for shaker table use
  • Vented or non-vented screw closure NOT autoclavable
  • Autoclavable flask
  • Sterile
Certifications
Certified sterile at 10-6 SAL USP Class VI
Specifications
Specifications
8.45 oz.
250 mL
For suspension cell culture, media preparation and storage
Autoclavable
Sterile
Baffled
Polypropylene
Polycarbonate
0.22μm PTFE pore
Yes
Class VI
Vented