missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Stack-Rack Space-Saver Tips: 101-1000μL
Increase limited bench space with Stack Racks
Description
Increase limited bench space with Stack Racks
- Racks nest together with each rack serving as a lid for the rack below
- Rack packs contain five racks of tips, with a base and dust cover
- Available in 5 × 100 formats
- Certified by independent laboratory
- Nonsterile; RNase, DNase, DNA, and pyrogen free
Specifications
100 to 1000 μL | |
Natural | |
7cm | |
Rack | |
Universal Fit |
Non-sterile | |
2.75 in. | |
DNase, RNase, DNA, and Pyrogen Free | |
Universal |