Fisherbrand™ Stainless Steel Clamps
Excellent chemical resistance and overall strength.
Supplier:  Fisherbrand™ 80862765
 View more versions of this product
Catalog No. 02-217-001
Explore available promotions
Description
Description
Excellent chemical resistance and overall strength.
  • Made of durable stainless-steel with electro-polished finish
  • Autoclavable
  • Extension clamps are supplied with vinyl and fiberglass prong covers for high temperature applications. Clamps secure various lab apparatus such as jointed glassware, columns, flask, and tubes.
  • S-Connector is ideal for connecting two perpendicular rods at a 90° angle and features separate adjustable screws for each rod
  • Lab-Foot Frame is designed for mounting lab-frames permanently to bench tops, fume hoods and floors
  • Jumbo Holder secures clamps to lab frames
  • Hook Connectors are simple, versatile, easy to use and allow one handed assembly of two components with one adjustment screw
Specifications
Specifications
1 Ea.
Stainless Steel
Jointed Glassware, Columns, Flask, Tube
9.02 in.
Medium
2.72 in.
Clamp
Vinyl and Fiberglass sleeves prong covers
Clamps secure various lab apparatus such as jointed Glassware, columns, flask and tubes
22.91 cm
6.9 cm
3-Prong