Fisherbrand™ Stainless Steel Clamps
Excellent chemical resistance and overall strength.
Supplier: Fisherbrand™ 80862767
Description
Excellent chemical resistance and overall strength.
- Made of durable stainless-steel with electro-polished finish
- Autoclavable
- Extension clamps are supplied with vinyl and fiberglass prong covers for high temperature applications. Clamps secure various lab apparatus such as jointed glassware, columns, flask, and tubes.
- S-Connector is ideal for connecting two perpendicular rods at a 90° angle and features separate adjustable screws for each rod
- Lab-Foot Frame is designed for mounting lab-frames permanently to bench tops, fume hoods and floors
- Jumbo Holder secures clamps to lab frames
- Hook Connectors are simple, versatile, easy to use and allow one handed assembly of two components with one adjustment screw
Specifications
1 Ea. | |
Stainless Steel | |
Ideal for connecting two perpendicular rods at a 90° angle and features separate adjustable screws for each rod | |
0.51 in. |
S-Connector | |
Connecting Two Perpendicular Rods at a 90 Deg. Angle and Features Separate Adjustable Screws for Each Rod | |
1.29 cm |