Fisherbrand™ Stainless Steel Clamps
Excellent chemical resistance and overall strength.
Supplier:  Fisherbrand™ 80862768
 View more versions of this product
Catalog No. 02-217-004
Explore available promotions
Description
Description
Excellent chemical resistance and overall strength.
  • Made of durable stainless-steel with electro-polished finish
  • Autoclavable
  • Extension clamps are supplied with vinyl and fiberglass prong covers for high temperature applications. Clamps secure various lab apparatus such as jointed glassware, columns, flask, and tubes.
  • S-Connector is ideal for connecting two perpendicular rods at a 90° angle and features separate adjustable screws for each rod
  • Lab-Foot Frame is designed for mounting lab-frames permanently to bench tops, fume hoods and floors
  • Jumbo Holder secures clamps to lab frames
  • Hook Connectors are simple, versatile, easy to use and allow one handed assembly of two components with one adjustment screw
Specifications
Specifications
1 Ea.
Stainless Steel
For mounting lab frames permanently to bench tops, fume hoods and floors
0.51 in.
Lab Frame Foot
0.75 in. Stainless Steel mounting screws
1.29 cm