Fisherbrand™ Steel Wire Handled Brushes Edge
Edge
Fisherbrand™ Steel Wire Handled Brushes feature stiff, white nylon bristles for cleaning test tubes.
 View more versions of this product
Catalog No. 03-572-5
Edge
Explore available promotions
Description
Description
Fisherbrand™ Stainless-Steel Handled Brushes feature stiff, white nylon bristles for cleaning test tubes.
  • Bristles are chemical-resistant
  • Radial tufted end
  • Stainless-steel handle is rustproof, corrosion resistant
Specifications
Specifications
Test Tube Brush
Radial, Tufted End
Radial Tufted End
1.37 in.
For cleaning test tubes
4 in.
10 cm
Nylon
Galvanized steel wire
White
3.5 cm
11 in.
27.94 cm
Small