Fisherbrand™ Superfrost™ Plus Microscope Slides Edge
Edge
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears.
 View more versions of this product
Catalog No. 22-037-246
Edge
Explore available promotions
Description
Description
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regular Superfrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Chemical-resistant white marking surface
  • Size: 25 × 75mm
Specifications
Specifications
Microscope Slide
Yes
25 mm
Clipped
Glass
75 mm
White
2 Packs of 72