Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Perfect for cell growth.
 View more versions of this product
Catalog No. FB012934
Explore available promotions
Description
Description
  • Flat, free flask surface
  • Striation free to maximize usable growth area
  • Used in both open, closed systems
  • Angled neck design offers good pipet and cell scraper access
  • Upper triangular, wider base shape provides stability
  • Protruding ridge on back side of flask makes it easy for stacking
  • Special area near neck for easy mark writing
  • Both flask sides have engraved graduation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Warranty
Three years
Specifications
Specifications
CE
Natural
Cell growth
200 Flasks
Tissue Culture Flask
Flat
Sterile
Plug Seal
Blue
25 mL
Angled
10 Flasks
Surface-treated
Yes
Flat
12.5 cm2