missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap

Perfect for cell growth.

$215.72 - $310.17

Specifications

Certifications/Compliance CE
Closure Color Blue
Color Natural
For Use With (Application) Cell growth
Neck Style Angled
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Culture Area Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Culture Area Price Quantity  
FB012934
View Documents
N/A 25 mL 12.5 cm2
Case of 200 Each for $288.83
 
FB012936
View Documents
N/A 50 mL 25 cm2
Case of 200 Each for $302.38
 
FB012938
View Documents
N/A 250 mL 75 cm2
Case of 100 Each for $310.17
 
FB012940
View Documents
N/A 600 mL 182 cm2
Case of 40 Each for $215.72
 
FB012942
View Documents
N/A 850 mL 300 cm2
Case of 18 Each for $225.97
 
Description

Description

  • Flat, free flask surface
  • Striation free to maximize usable growth area
  • Used in both open, closed systems
  • Angled neck design offers good pipet and cell scraper access
  • Upper triangular, wider base shape provides stability
  • Protruding ridge on back side of flask makes it easy for stacking
  • Special area near neck for easy mark writing
  • Both flask sides have engraved graduation
  • Sterilized by gamma irradiation
  • Non-pyrogenic

Warranty

Three years

Specifications

Specifications

CE
Natural
Angled
Surface-treated
Yes
Flat
Blue
Cell growth
Tissue Culture Flask
Flat
Sterile
Plug Seal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'