Fisherbrand™ Surface Treated Tissue Culture Dishes
Containers for cell growth, aids in sample preparation, pretreatment, storage.
 View more versions of this product
Catalog No. FB012922
Explore available promotions
Description
Description
  • Flat bottom uniform wall thickness to ensure distortion free bottom
  • Smooth, free surface
  • Less striation to maximize usable area for growth
  • Rim on upper side of lid mates with dish brim for easy, secure stack
  • Lids with chimbs to shape vents
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Warranty
Three years
Specifications
Specifications
CE
Cell and tissue culture dishes
36.3 cm2
Sterile
Tissue culture, storage
10 Plates
70 mm
Yes
Flat
Virgin Polystyrene
Standard
600 Plates
15.0 x 73.5 mm