Fisherbrand™ Tissue Path™ IV Tissue Cassettes
Contain no jagged edges or uneven surfaces
Supplier: Fisherbrand™ 58411
Description
- Unique, two-piece molded hinge ensures specimen safety and facilitates easy removal of the cassette cover from the base during the embedding procedure
- Cover and base can be disassembled and reassembled with ease
- Cassettes fit all sizes of popular base molds, metal and plastic
- Three-sided writing surface assures positive identification
- Slotted design ensures thorough and rapid exchange of processing fluids
- Resistant to solvents, decalcifying solutions, and compatible with microwave techniques
Order Info
- Supplied in cases of four handy dispenser boxes, 250 per box
Specifications
Slots | |
Yellow | |
Plastic | |
Tissue Cassette |